Pipette controller, Pi-Pump
Pipette controller Pi-Pump, green, for 10 ml pipettes
N° de catalogue 612-1926
Fournisseur: KARTELL
Jump to:
Product Details & Documents
Pipetting aid for all plastic and glass pipettes.
- Suction and dispensing achieved by rotating the wheel
- Liquid evacuated by pressing the lever
Documents
Specifications
- Description:Pipette controller Pi-Pump, green, for 10 ml pipettes
- Cat. No.:612-1926