Order Entry
Belgium
Orders LinkContactUsLinkComponent
TLC developing chambers

TLC developing chambers

TLC separating chamber, for 50×200 mm plates, with cap, 60×230 mm
Catalog # 552-5272
Supplier:   witeg Labortechnik
TLC developing chambers
Catalog # 552-5272
Supplier:  witeg Labortechnik
TLC developing chambers
TLC developing chambers

TLC separating chamber, for 50×200 mm plates, with cap, 60×230 mm

Price
79,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Round shaped chambers made of borosilicate glass.

Specifications
  • Description:
    TLC separating chamber, for 50×200 mm plates, with cap
  • Seq:
    01
  • Ø×H:
    60×230 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.