Order Entry
Belgium
Orders LinkContactUsLinkComponent
WHEATON® Serum Vials

WHEATON® Serum Vials

int.Ø: 13 mm, ext.Ø: 20 mm (Mouth), 20,00 ml, Flat bottom, 30,5×58 mm, Glass, Clear, No
Catalog # 548-2014
Supplier:   DWK Life Sciences
WHEATON® Serum Vials
Catalog # 548-2014
Supplier:  DWK Life Sciences
WHEATON® Serum Vials
WHEATON® Serum Vials

int.Ø: 13 mm, ext.Ø: 20 mm (Mouth), 20,00 ml, Flat bottom, 30,5×58 mm, Glass, Clear, No

Price
220,00 €
Log in for availability
1 * 120 ST
Product Details & Documents

These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320'  borosilicate glass.

  • Blowback feature in the neck and a specially designed bottom radius for added strength
  • These features are important for freeze-drying applications
  • Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination

Certifications: Meets ASTM Type 1, Class A/B and USP Type 1 standards.

Delivery information: Vials are supplied without closures.

Specifications
  • Cap included:
    No
  • Ø×H:
    30,5×58 mm
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Description:
    int.Ø: 13 mm, ext.Ø: 20 mm (Mouth)
  • Material:
    Glass
  • Seq:
    11
  • Base:
    Flat bottom
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.