Orders LinkContactUsLinkComponent
Jars, with lid
Jars, with lid
50 mm, 30 mm
Catalog # 525-1364
Jars, with lid
Catalog # 525-1364
Jars, with lid
Jars, with lid
50 mm, 30 mm
Price
13,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Specifications
  • Ext.Ø:
    50 mm
  • Height:
    30 mm