ContactUsLinkComponent
P1-H70 Bridge Circulator, 30C to 250C Working Temperature Range, 2kW Heater Capacity, 320 to 800mm Bath Bridge Width, 1 * 1 items
Catalog # 461-5101
Supplier:  HAAKE
P1-H70 Bridge Circulator, 30C to 250C Working Temperature Range, 2kW Heater Capacity, 320 to 800mm Bath Bridge Width, 1 * 1 items
Catalog # 461-5101
Supplier:  HAAKE
Frequently Bought Together