Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bath fluids

Therm 160, 60...160 °C, >273 °C, >263 °C, Brown, 10 l
Catalog # 461-2861
Supplier:   LAUDA
Bath fluids
Catalog # 461-2861
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Therm 160, 60...160 °C, >273 °C, >263 °C, Brown, 10 l

CAS Number:  
Price
669,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    10 l
  • Colour:
    Brown
  • Fire point:
    >273 °C
  • Flash point:
    >263 °C
  • Temperature range:
    60...160 °C
  • Type:
    Therm 160

Specifications