Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bath fluids, Haake

Bath fluids, Haake

Sil 100, –75...+75 °C, >100 °C, 57 °C, 3 cP, Colourless, 5 L
Catalog # 461-0010
Supplier:   THERMO ELECTRON LED
Bath fluids, Haake
Catalog # 461-0010
CAS Number:  
Bath fluids, Haake

Sil 100, –75...+75 °C, >100 °C, 57 °C, 3 cP, Colourless, 5 L

CAS Number:  
Price
649,00 €
Log in for availability
1 * 5 L
Product Details & Documents

These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.

  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency

The oils are not suitable for contact with silicone tubing.

Documents

Specifications

Specifications

  • Pk:
    5 L
  • Colour:
    Colourless
  • Fire point:
    >100 °C
  • Flash point:
    57 °C
  • Temperature range:
    –75...+75 °C
  • Type:
    Sil 100
  • Viscosity:
    3 cP

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.