Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pestles

28 mm, 125 mm, 410-9121, 410-9111
  410-9137
Pestles
  410-9137
Pestles
Pestles

28 mm, 125 mm, 410-9121, 410-9111

 
41,20 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9121, 410-9111
  • Head Ø:
    28 mm
  • Length:
    125 mm
  • Seq:
    02