Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
Black, FLUOTRAC™ 600 high binding, sterile, 40 - 340 µl, 96, +, 392 µl
Catalog # 391-3629
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 96-well
Catalog # 391-3629
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 96-well
Microplates, 96-well
Black, FLUOTRAC™ 600 high binding, sterile, 40 - 340 µl, 96, +, 392 µl
Price
115,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications
  • No. of wells:
    96
  • Recommended working volume:
    40 - 340 µl
  • Sterile:
    +
  • Description:
    Black, FLUOTRAC™ 600 high binding, sterile
  • Seq:
    13
  • Total volume:
    392 µl
  • Cat. No.:
    391-3629
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact