Orders LinkContactUsLinkComponent
Adhesive seals
Adhesive seals
EASYseal adhesive film, non sterile, 80×140 mm, Transparent, -
Catalog # 736-0127
Supplier:   Greiner Bio-One
Adhesive seals
Catalog # 736-0127
Supplier:  Greiner Bio-One
Adhesive seals
Adhesive seals
EASYseal adhesive film, non sterile, 80×140 mm, Transparent, -
Price
51,40 €
Log in for availability
1 * 100 ST
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
  • Free from RNase, DNase and human DNA
  • Non pyrogenic
Specifications
  • Optical property:
    Transparent
  • Sterile:
    -
  • Description:
    EASYseal adhesive film, non sterile, 80×140 mm
  • Seq:
    02