Microplates, 96-well
96-well microplate, F-bottom (chimney well), MICROLON® 600, 25 - 340 µl, 392 µl
Catalog # 391-3390
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifications
- Description:96-well microplate, F-bottom (chimney well), MICROLON® 600
- Seq:08
- Working volume:25 - 340 µl
- Total volume:392 µl
- Cat. No.:391-3390
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact