Clamps, universal
Stainless steel, 0 - 80 mm
Catalog # 241-7311
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
The fingers of the clamps have cork inserts to protect the clamped items.
- Without bosshead for fixing to stand
- With T-screws for rapid easy clamping
Specifications
- Spanning width:0 - 80 mm
- Material:Stainless steel