0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Retort Rods
Stand rods
Stand rods
12 mm, 1000 mm
241-7143
:
USBECK Laborgeräte
Stand rods
241-7143
:
USBECK Laborgeräte
Stand rods
12 mm, 1000 mm
24,30 €
1 * 1 ST
-
+
Stainless steel.
Robust, very stable
Excellent chemical resistance
Ø:
12 mm
Length:
1000 mm
Seq:
14