Order Entry
Belgium
Orders LinkContactUsLinkComponent
Squared bossheads

Squared bossheads

Aluminium alloy, 12 - 13 × 12 - 13 mm
Catalog # 241-3172
Supplier:   BOCHEM
Squared bossheads
Catalog # 241-3172
Supplier:  BOCHEM
Squared bossheads
Squared bossheads

Aluminium alloy, 12 - 13 × 12 - 13 mm

Price
22,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Squared bossheads with hexagon socket screws.

  • Made from 18/10 stainless steel or aluminium (laboral)
  • Angle: 90°
Specifications
  • Material:
    Aluminium alloy
  • Seq:
    02
  • For rod Ø:
    12 - 13 × 12 - 13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.