Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electronic cell counters

Electronic cell counters

Electronic cell counter, AC-10, 10
Catalog # 233-5394
Electronic cell counters
Catalog # 233-5394
Electronic cell counters
Electronic cell counters

Electronic cell counter, AC-10, 10

Price
842,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The electronic memory devices specify blood counting.

  • Power supply operation
  • Rechargeable battery operation
  • Software (download) with computer analysis
  • Big TFT color display
  • Storage of last counting on device

The microscope image can be observed permanently, the numbers can be typed in easily.

Colour: Blue

Delivery information: Every device comes with an instruction manual.

Specifications
  • Model:
    AC-10
  • Description:
    Electronic cell counter
  • Seq:
    001
  • Array:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.