Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 160 mm
Catalog # 233-1222
Supplier:   RSG
Laboratory scissors, straight
Catalog # 233-1222
Supplier:  RSG
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 160 mm
Price
9,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Stainless steel, INOX 4021.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Spécifications
  • Type:
    Sharp/blunt
  • Length:
    160 mm