Laboratory scissors, straight
Sharp/sharp, 145 mm
Catalog # 233-1214
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel, INOX 4021.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Designed for rough handling
Documents
RSGA103.130_TDS_EN RSGA105.130_TDS_EN
Datasheet
Specifications
- Type:Sharp/sharp
- Length:145 mm