Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, pointed, 160 mm
Catalog # 232-2125
Supplier:   RSG
Forceps
Catalog # 232-2125
Supplier:  RSG
Forceps
Forceps
Straight, pointed, 160 mm
Price
10,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
Documents
Specifications
  • Type:
    Straight, pointed
  • Length:
    160 mm