Micro forceps
No. 5-45, laterally angled (45°), very fine tips, 110 mm
Catalog # 232-1922
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Stainless steel.
Specifications
- Type:No. 5-45, laterally angled (45°), very fine tips
- Length:110 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



