Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

210 mm, 5×7 mm
Catalog # 231-2164
Supplier:   HAMMACHER KARL
Spoon spatulas, micro
Catalog # 231-2164
Supplier:  HAMMACHER KARL
Spoon spatulas, micro
Spoon spatulas, micro

210 mm, 5×7 mm

Price
13,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Double-ended spatulas with one spoon end and one spatula end. Made of stainless steel.

Specifications
  • Length:
    210 mm
  • Seq:
    04
  • Spoon size:
    5×7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.