Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions, Specpure®

1,68, ±0,01, HCl/KCl, Colourless, 25 °C, Glass bottle, 500 mL
Catalog # 42415.AP
pH buffer solutions, Specpure®
Catalog # 42415.AP
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

1,68, ±0,01, HCl/KCl, Colourless, 25 °C, Glass bottle, 500 mL

CAS Number:  
Price
35,90 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • pH buffer type:
    HCl/KCl
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • pH accuracy:
    ±0,01
  • pH value:
    1,68

Specifications