Fused quartz microscope slides
Microscope slides, Fused quartz, 26×26 mm
These fused quartz slides are made of fusing naturally occurring crystalline silica.
- High chemical purity, high corrosion resistance, high melting point
- Low coefficient of thermal expansion with excellent electrical insulation
- Optical transmission from ultra-violet to infrared
- Description:Microscope slides
- L×W:26×26 mm
- Material:Fused quartz
- Seq:04