Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®

Sample vials in Lab File®, WHEATON®

16,00 ml, 21×70 mm, Clear, No, 18-400
Catalog # 218-2225
Supplier:   DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Catalog # 218-2225
Supplier:  DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

16,00 ml, 21×70 mm, Clear, No, 18-400

Price
232,00 €
Log in for availability
1 * 200 ST
Product Details & Documents

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

Ordering information: Without cap.

Specifications
  • Cap included:
    No
  • Cap Size:
    18-400
  • Ø×H:
    21×70 mm
  • Capacity:
    16,00 ml
  • Colour:
    Clear
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.