Sample vials in Lab File®, WHEATON®
12,00 ml, 19×65 mm, Clear, No, 15-425
Catalog # 218-2224
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.
Ordering information: Without cap.
Documents
Specifications
- Cap included:No
- Cap Size:15-425
- Ø×H:19×65 mm
- Capacity:12,00 ml
- Colour:Clear
- Seq:08



