Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample vials in Lab File®, WHEATON®

Sample vials in Lab File®, WHEATON®

40,00 ml, 28×95 mm, Clear, No, 24-400
Catalog # 218-2203
Supplier:   DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Catalog # 218-2203
Supplier:  DWK Life Sciences
Sample vials in Lab File®, WHEATON®
Sample vials in Lab File®, WHEATON®

40,00 ml, 28×95 mm, Clear, No, 24-400

Price
437,00 €
Connexion Pour la disponibilité
1 * 200 ST
Product Details & Documents

For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.

Ordering information: Without cap.

Spécifications
  • Cap included:
    No
  • Cap Size:
    24-400
  • Ø×H:
    28×95 mm
  • Capacity:
    40,00 ml
  • Colour:
    Clear
  • Seq:
    12