Orders LinkContactUsLinkComponent
Wire baskets, square
Wire baskets, square
300×300×300 mm
Catalog # 216-7512
Supplier:   DELTALAB
Wire baskets, square
Catalog # 216-7512
Supplier:  DELTALAB
Wire baskets, square
Wire baskets, square
300×300×300 mm
Price
101,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Square baskets, made of stainless steel wire, with folding handles.
  • Features a strong woven mesh base
  • Good chemical resistance
  • Good temperature resistance
  • Comes without lid (to order baskets with lid please contact our customer service
Documents
Specifications
  • Seq:
    05
  • W×D×H:
    300×300×300 mm