Orders LinkContactUsLinkComponent
Autoclavable baskets
Autoclavable baskets
470×300×200 mm
N° de catalogue 216-7491
Fournisseur:   DELTALAB
Autoclavable baskets
N° de catalogue 216-7491
Fournisseur:  DELTALAB
Autoclavable baskets
Autoclavable baskets
470×300×200 mm
Prix
260,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of stainless steel wire.
  • Stackable
  • Good chemical resistance
  • Excellent temperature resistance
Baskets for autoclaves and carrying.
Documents
Specifications
  • Seq:
    01
  • W×D×H:
    470×300×200 mm