Orders LinkContactUsLinkComponent
Autoclavable baskets
Autoclavable baskets
570×300×150 mm
Catalog # 216-7490
Supplier:   DELTALAB
Autoclavable baskets
Catalog # 216-7490
Supplier:  DELTALAB
Autoclavable baskets
Autoclavable baskets
570×300×150 mm
Price
142,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of stainless steel wire.
  • Stackable
  • Good chemical resistance
  • Excellent temperature resistance
Baskets for autoclaves and carrying.
Documents
Specifications
  • Seq:
    02
  • W×D×H:
    570×300×150 mm