Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Test tubes

5 ml, 74,2 mm, 11,8 mm, graduated, 10,3 mm
Catalog # 212-3321
Supplier:   DELTALAB
Test tubes
Catalog # 212-3321
Supplier:  DELTALAB
Test tubes
Test tubes

5 ml, 74,2 mm, 11,8 mm, graduated, 10,3 mm

Price
39,10 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Polypropylene (PP), ultra clear.

  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable

Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.

Specifications
  • Ext.Ø:
    11,8 mm
  • Ø int.:
    10,3 mm
  • Version:
    graduated
  • Capacity:
    5 ml
  • Length:
    74,2 mm
  • Seq:
    03