Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test tubes

Test tubes

3 ml, 54,3 mm, 11,6 mm, ungraduated, 9,9 mm
Catalog # 212-3320
Supplier:   DELTALAB
Test tubes
Catalog # 212-3320
Supplier:  DELTALAB
Test tubes
Test tubes

3 ml, 54,3 mm, 11,6 mm, ungraduated, 9,9 mm

Price
92,90 €
Log in for availability
1 * 2.000 ST
Product Details & Documents

Polypropylene (PP), ultra clear.

  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable

Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.

Specifications
  • Ext.Ø:
    11,6 mm
  • Ø int.:
    9,9 mm
  • Version:
    ungraduated
  • Capacity:
    3 ml
  • Length:
    54,3 mm
  • Seq:
    01