Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stools

Stools

Roll stool to step on, plastic, Ø 300 mm, 420 mm
Catalog # 139-1175
Supplier:   BOCHEM
Stools
Catalog # 139-1175
Supplier:  BOCHEM
Stools
Stools

Roll stool to step on, plastic, Ø 300 mm, 420 mm

Price
205,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Kickstool with GS safety control.

Specifications
  • Description:
    Roll stool to step on, plastic, Ø 300 mm
  • Height:
    420 mm
  • Seq:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.