Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic (≥79% Bi₂O₃)

Bismuth(III) nitrate basic (≥79% Bi₂O₃)

Glass bottle for solids, 100 g
Catalog # 40476.22
Oxydant
Irritant
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Catalog # 40476.22
CAS Number:  1304-85-4
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Bismuth(III) nitrate basic (≥79% Bi₂O₃)

Glass bottle for solids, 100 g

CAS Number:  1304-85-4
Price
82,80 €
Connexion Pour la disponibilité
1 * 100 g
Product Details & Documents

CAS No.: 1304-85-4 Molecular Formula: Bi5O(OH)9(NO3)4 (approximate) Formula Weight: 1461.98 Physical Form: Powder Density: 6.04 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 260° dec. Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl Merck Reference: 14,1284 UN No.: 1477 MDL No.: MFCD00064844

Ceramic glazes and enamel fluxes

Synonymes:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • Numéro CAS:1304-85-4
  • Formule:Bi₅O(OH)₉(NO₃)₄
  • Numéro MDL:MFCD00064844
  • Poids moléculaire:1461.99 g/mol
  • Index Merck:14,1284
  • Point de fusion:260 °C (dec.)
  • Densité:6.04
  • Température de stockage:Ambient
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Spécifications

Spécifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Spécifications