Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Catalog # 40476.14
Supplier: Thermo Fisher Scientific
CAS Number: 1304-85-4
Jump to:
Product Details & Documents
CAS No.: 1304-85-4 Molecular Formula: Bi5O(OH)9(NO3)4 (approximate) Formula Weight: 1461.98 Physical Form: Powder Density: 6.04 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 260° dec. Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl Merck Reference: 14,1284 UN No.: 1477 MDL No.: MFCD00064844
Ceramic glazes and enamel fluxes
Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
- CAS Number:1304-85-4
- Formula:Bi₅O(OH)₉(NO₃)₄
- MDL Number:MFCD00064844
- Molecular Weight:1461.99 g/mol
- Merck Index:14,1284
- Melting Pt:260 °C (dec.)
- Density:6.04
- Storage Temperature:Ambient
- EINECS:215-136-8
- UN:1477
- ADR:5.1,II
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



