pH buffer solutions, Specpure®
7,00, ±0,01, Phosphate, Green, 25 °C, Glass bottle, 1 L
Fournisseur: Thermo Fisher Scientific
pH buffer solutions, Specpure®
N° de catalogue 38711.K2
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Jump to:
Product Details & Documents
- NIST traceable
- Individual expiration date stickers
Documents
Specifications
Specifications
- Pack type:Glass bottle
- pH buffer type:Phosphate
- Pk:1 L
- Reference temperature:25 °C
- Colour:Green
- pH accuracy:±0,01
- pH value:7,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



