Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, Specpure®

pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Colourless, 25 °C, Glass bottle, 4 L
Catalog # 38710.K7
pH buffer solutions, Specpure®
Catalog # 38710.K7
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Colourless, 25 °C, Glass bottle, 4 L

CAS Number:  
Price
106,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • pH buffer type:
    Carbonate
  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • pH accuracy:
    ±0,01
  • pH value:
    10,00
  • Cat. No.:
    38710.K7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.