pH buffer solutions, Specpure®
10,00, ±0,01, Carbonate, Colourless, 25 °C, Glass bottle, 1 L
- NIST traceable
- Individual expiration date stickers
- Pack type:Glass bottle
- pH buffer type:Carbonate
- Pk:1 L
- Reference temperature:25 °C
- Colour:Colourless
- pH accuracy:±0,01
- pH value:10,00



