Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol ≥98%

D-(-)-Sorbitol ≥98%

Glass bottle for solids, 500 g
Catalog # 36404.36
D-(-)-Sorbitol ≥98%
Catalog # 36404.36
CAS Number:  50-70-4
D-(-)-Sorbitol ≥98%
D-(-)-Sorbitol ≥98%

Glass bottle for solids, 500 g

CAS Number:  50-70-4
Price
49,40 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 50-70-4 Molecular Formula: C6H14O6 Formula Weight: 182.17 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 98-100° Solubility: Soluble in water, methanol, acetone, DMF Beilstein Reference: 1721899 Merck Reference: 14,8725 MDL No.: MFCD00004708

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,8725
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:98…100 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications