L(+)-Ascorbic acid 99+% ACS
Glass bottle for solids, 100 g
Fournisseur: Thermo Fisher Scientific
L(+)-Ascorbic acid 99+% ACS
N° de catalogue 36237.22
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 50-81-7
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:14,830
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:∼ 191 °C (dec.)
- Density:1.65
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



