Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid 99+% ACS

Glass bottle for solids, 25 g
N° de catalogue 36237.14
Fournisseur:   Thermo Fisher Scientific
L(+)-Ascorbic acid 99+% ACS
N° de catalogue 36237.14
Numéro CAS:  50-81-7
L(+)-Ascorbic acid 99+% ACS
L(+)-Ascorbic acid 99+% ACS

Glass bottle for solids, 25 g

Numéro CAS:  50-81-7
Prix
40,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:∼ 191 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications