Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23

Rods, ceramic, round, solid, AL23

5,0 mm, 250 mm
Catalog # 32931.DT
Rods, ceramic, round, solid, AL23
Catalog # 32931.DT
Rods, ceramic, round, solid, AL23

5,0 mm, 250 mm

Price
77,80 €
Log in for availability
1 * 250 mm
Product Details & Documents

Pure Al₂O₃ Aluminium oxide, AL23.

Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.

Specifications
  • Ext.Ø:
    5,0 mm
  • Length:
    250 mm
  • Seq:
    43
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.