Orders LinkContactUsLinkComponent
Graphite electrodes, counter
Graphite electrodes, counter
Graphite electrode, counter-flat top, 3,05×38,10 mm
Catalog # 14754.KN
Graphite electrodes, counter
Catalog # 14754.KN
Graphite electrodes, counter
Graphite electrodes, counter
Graphite electrode, counter-flat top, 3,05×38,10 mm
Price
434,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Graphite electrodes are used as homogeneous catalysts.
  • AGKSP grade
  • Ultra F purity
  • Light sensitive and hygroscopic
  • Incompatible with oxidising agents
  • Applications include piston rings, thrust bearings, journal bearings and vanes
These are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.
Specifications
  • Ø×L:
    3,05×38,10 mm
  • Description:
    Graphite electrode, counter-flat top
  • Seq:
    01
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.