Lithium hydroxide, anhydrous 98%
Plastic bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Lithium hydroxide, anhydrous 98%
Catalog # 13407.22
Supplier: Thermo Fisher Scientific
CAS Number: 1310-65-2
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: -4+14 Mesh Density: 2.54 Appearance: White Odour: Odourless pH: 14 Melting Point: 462° Boiling Point: 924° Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095
- CAS Number:1310-65-2
- Formula:LiOH
- MDL Number:MFCD00011095
- Molecular Weight:23.95 g/mol
- Merck Index:14,5534
- Boiling Pt:924 °C
- Melting Pt:462 °C
- Density:2.54
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



