Precision probes, T
TC T-type robust food penetration probe with reinforced handle, −50…+350 °C, ±0,2 (−20…+70);
±1,0 °C, 5×115 mm
±1,0 °C, 5×115 mm
Catalog # 620-3658
Supplier: TESTO
Jump to:
Product Details & Documents
Specifications
- Range:−50…+350 °C
- Description:TC T-type robust food penetration probe with reinforced handle
- Accuracy:±0,2 (−20…+70);
±1,0 °C - Ø×L:5×115 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



