Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phosphate standard solutions

Phosphate standard solutions

Phosphate standard solution, pack of 16×10 ml voluette ampoules (NIST), 50 mg/l as PO4, 16
Catalog # 705-0076
Supplier:   Hach
Phosphate standard solutions
Catalog # 705-0076
Supplier:  Hach
CAS Number:  
Phosphate standard solutions
Phosphate standard solutions

Phosphate standard solution, pack of 16×10 ml voluette ampoules (NIST), 50 mg/l as PO4, 16

CAS Number:  
Price
70,10 €
Log in for availability
1 * 16 ST
Product Details & Documents

Standard solution for accuracy checks in phosphate (phosphorus) determinations.

Specifications

Specifications

  • Concentration:
    50 mg/l as PO<sub>4</sub>
  • Pk:
    16
  • Description:
    Phosphate standard solution, pack of 16×10 ml voluette ampoules (NIST)

Specifications