Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions

Type L4798, 4,01/6,87/9,18, ±0,010, 25 °C, 60 Ampoul
Catalog # 662-1429
Supplier:   SI Analytics
Buffer solutions
Catalog # 662-1429
Supplier:  SI Analytics
CAS Number:  
Buffer solutions
Buffer solutions

Type L4798, 4,01/6,87/9,18, ±0,010, 25 °C, 60 Ampoul

CAS Number:  
Price
265,00 €
Log in for availability
1 * 60 Ampul
Product Details & Documents

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

 : According to DIN 19 266.

Specifications

Specifications

  • Pk:
    60 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Type L4798
  • pH accuracy:
    ±0,010
  • pH value:
    4,01/6,87/9,18

Specifications