Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Disodium tetraborate decahydrate, 9,184 (25°C), Plastic bottle, 5×100 ml, 5
Supplier: Merck
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Catalog # 1.07203.0105
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Materials for the precise calibration and monitoring of pH instruments.
- Traceable to NIST primary standard and PTB
- Maximum deviation ±0,003 pH units
- Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
Documents
Certificates
Search by Lot NumberCertipur
Product Literature
MERC1.07203.0105_TDS_EN
Datasheet
ProductPage_101964 ProductPage_101964 ProductPage_101965
Product specification
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:5
- Description:Disodium tetraborate decahydrate
- Pk Info:5×100 ml
- pH value:9,184 (25°C)
- Cat. No.:1.07203.0105
- M-Clarity quality level:MQ300
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



