Cavity slides
Cavity slides with PTFE coating, Black, 8, 6 mm
N° de catalogue 631-0465
Fournisseur: Epredia
Jump to:
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).
Documents
Specifications
- No. of wells:8
- Colour:Black
- Description:Cavity slides with PTFE coating
- Seq:020
- Well Ø:6 mm