Orders LinkContactUsLinkComponent
Multiple or single cuvette segments
Multiple or single cuvette segments
Cuvettes in pre-racked segments (15), PMMA
Catalog # KART1500
Supplier:   KARTELL
Multiple or single cuvette segments
Catalog # KART1500
Supplier:  KARTELL
Multiple or single cuvette segments
Multiple or single cuvette segments
Cuvettes in pre-racked segments (15), PMMA
Price
858,00 €
Log in for availability
1 * 450 ST
Product Details & Documents
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Packaging: In cases of bulk segments (450) or 30 pre-racked segments (15).
Specifications
  • Description:
    Cuvettes in pre-racked segments (15)
  • Material:
    PMMA
  • Seq:
    002