Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pestles

Pestles

40 mm, 160 mm, 410-0084
Catalog # 410-0087
Supplier:   VITLAB
Pestles
Catalog # 410-0087
Supplier:  VITLAB
Pestles
Pestles

40 mm, 160 mm, 410-0084

Price
51,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Melamine-formaldehyde, white.

Specifications
  • For:
    410-0084
  • Head Ø:
    40 mm
  • Length:
    160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.