Order Entry
Belgium
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

250 ml, 95 mm, 77 mm, 25 ml
Catalog # 213-1710
Supplier:   VITLAB
Beakers, low form
Catalog # 213-1710
Supplier:  VITLAB
Beakers, low form
Beakers, low form

250 ml, 95 mm, 77 mm, 25 ml

Price
3,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, highly transparent.

  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning

Ideal for general laboratory use. With spout and exceptionally easy-to-read embossed blue graduations.

Food-safe products acc. to EC Directive No. 10/2011.

Certifications: ISO 7056 - 10 ml/150 ml/400 ml/600 ml/3000 ml in addition to ISO 7056)

Specifications
  • Ext.Ø:
    77 mm
  • Graduation Interval:
    25 ml
  • Capacity:
    250 ml
  • Height:
    95 mm
  • Seq:
    06
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.